Tisk…

Copper(II) acetylacetonate ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

A14920.0B A14920.18 A14920.22
A14920.0BEA 10130 CZK
A14920.0B A14920.18 A14920.22
Copper(II) acetylacetonate ≥98%
Copper(II) acetylacetonate

Purity: 98% CAS No.: 13395-16-9 Molecular Formula: C10H14CuO4 Formula Weight: 261.76 Odour: Weak Melting Point: ca 286° dec. Boiling Point: 160°/10mm Beilstein Reference: 4157957 MDL No.: MFCD00000016

Vzorec: C₁₀H₁₄CuO₄
MW: 261.76
Bod varu: 160 °C (10 mmHg)
Bod tání: ∼ 286 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00000016
CAS číslo: 13395-16-9
EINECS: 236-477-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR