Copper(II) acetylacetonate, hydrate
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate
12535.14
12535.22
12535.36
12535.14EA
865
CZK
12535.14
12535.22
12535.36
Copper(II) acetylacetonate, hydrate
Copper(II) acetylacetonate
CAS No.: 13395-16-9
Molecular Formula: C10H14CuO4.xH2O
Formula Weight: 261.76anhy
Physical Form: Powder
Odour: Weak
Melting Point: 230°
MDL No.: MFCD00000016
Vzorec:
C₁₀H₁₄CuO₄ MW: 261.76 Bod tání: 230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000016 CAS číslo: 13395-16-9 EINECS: 236-477-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...