Copper(II) acetylacetonate 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate
110651000
110650250
110655000
ACRO110651000EA
1820
CZK
ACRO110651000
ACRO110650250
ACRO110655000
Copper(II) acetylacetonate 98%
Copper(II) acetylacetonate
Vzorec:
C₁₀H₁₄CuO₄ MW: 261.76 Bod varu: 160 °C (9.8 mmHg) Bod tání: 245 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00000016 CAS číslo: 13395-16-9 EINECS: 236-477-9 |
Výsledky specifikačního testu
Appearance | Blue to blue-grey Crystalline powder |
Infrared spectrum | Conforms |
Titration with Na2S2O3 | 97.5 to 102.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...