Tisk…

Danzylchlorid ~98%, yellow powder

Dodavatel: MP Biomedicals
Nebezpečí

Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

0210060305 0210060301
ICNA0210060305EA 0 CZK
ICNA0210060305 ICNA0210060301
Danzylchlorid ~98%, yellow powder
Danzylchlorid

Dansyl Chloride is a fluorogenic reagent for fluorescent N-terminal derivatisation of amino acids and peptides and detection by reverse phase HPLC. It is used in labeling proteins and amino acids.


Dansyl Chloride is a fluorogenic reagent for fluorescent N-terminal derivatisation of amino acids and peptides and detection by reverse phase HPLC. It is used in labeling proteins and amino acids.


Soluble in acetone (10% w/v - clear solution).

Vzorec: C₁₂H₁₂ClNO₂S
MW: 269,75 g/mol
Bod tání: 64…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS číslo: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,III
Merck Index: 13,02840

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR