Tisk…

6-Bromo-2-naphthol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

M-Clarity™ quality level = MQ100

B73406-100G B73406-25G
SIALB73406-100GEA 8520 CZK
SIALB73406-100G SIALB73406-25G
6-Bromo-2-naphthol, Sigma-Aldrich®
6-Bromo-2-naphthol
Vzorec: C₁₀H₇BrO
MW: 223,07 g/mol
Bod varu: 200…205 °C (20 mmHg)
Bod tání: 128 °C
MDL Number: MFCD00004081
CAS číslo: 15231-91-1
EINECS: 239-279-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR