Tisk…

6-Bromo-2-naphthol ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

A18535.18 A18535.09
A18535.18EA 3470 CZK
A18535.18 A18535.09
6-Bromo-2-naphthol ≥97%
6-Bromo-2-naphthol

Purity: 97% CAS No.: 15231-91-1 Molecular Formula: C10H7BrO Formula Weight: 223.08 Melting Point: 126-129° Beilstein Reference: 1100270

Vzorec: C₁₀H₇BrO
MW: 223,07 g/mol
Bod varu: 200…205 °C (20 mmHg)
Bod tání: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00004081
CAS číslo: 15231-91-1
EINECS: 239-279-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR