Oxid ceričitý, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Ceric oxide, Cerium dioxide
M-Clarity™ quality level = MQ100
700290-100G
700290-25G
SIAL700290-100GEA
11750
CZK
SIAL700290-100G
SIAL700290-25G
Oxid ceričitý, Sigma-Aldrich®
Oxid ceričitý
Vzorec:
CeO₂ MW: 172.11 Bod varu: 3468 Bod tání: 2000 Hustota: 7.13 |
MDL Number:
MFCD00010933 CAS číslo: 1306-38-3 EINECS: 215-150-4 Merck Index: 12,02035 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...