Tisk…

Praseodymium(III) nitrate hexahydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Praseodymium trinitrate hexahydrate

M-Clarity™ quality level = MQ200

205133-10G 205133-250G 205133-50G
SIAL205133-10GEA 2370 CZK
SIAL205133-10G SIAL205133-250G SIAL205133-50G
Praseodymium(III) nitrate hexahydrate, Sigma-Aldrich®
Praseodymium(III) nitrate hexahydrate
Vzorec: N₃O₉Pr·6H₂O
MW: 435,01 g/mol
Bod tání: 56 °C
Hustota: 2,233 g/cm³ (20 °C)
MDL Number: MFCD00149826
CAS číslo: 15878-77-0
EINECS: 233-796-5
UN: 1477
ADR: 5.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR