Lanthanum(III) perchlorate hexahydrate
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Lanthanum perchlorate hexahydrate
11641.14
11641.22
11641.14EA
1680
CZK
11641.14
11641.22
Lanthanum(III) perchlorate hexahydrate
Lanthanum(III) perchlorate hexahydrate
CAS No.: 14017-46-0
Molecular Formula: La(ClO4)3.6H2O
Formula Weight: 545.35 (437.26anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Odour: Odourless
UN No.: 1481
MDL No.: MFCD00016159
Vzorec:
Cl₃LaO₁₂·6H₂O MW: 545,35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016159 CAS číslo: 36907-37-6 EINECS: 237-835-7 UN: 1481 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...