Tisk…

Dimethyl trisulphide ≥98%

Dodavatel: Thermo Scientific
Varování

H32913.06 H32913.14
H32913.06EA 3230 CZK
H32913.06 H32913.14
Dimethyl trisulphide ≥98%
Dimethyl trisulphide

Purity: 98% CAS No.: 3658-80-8 Molecular Formula: C2H6S3 Formula Weight: 126.26 Flash Point: 52°(126°F) Refractive Index: 1.600 Odour: Stench Boiling Point: 165-175° UN No.: 1993 MDL No.: MFCD00039808

Vzorec: C₂H₆S₃
MW: 126,27 g/mol
Bod varu: 165…175 °C
Hustota: 1,203
Bod vzplanutí: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039808
CAS číslo: 3658-80-8
EINECS: 222-910-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR