Tisk…

Selenium trioxide, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Selenium dioxide

M-Clarity™ quality level = MQ200

204315-10G 204315-50G
SIAL204315-10GEA 4910 CZK
SIAL204315-10G SIAL204315-50G
Selenium trioxide, Sigma-Aldrich®
Selenium trioxide
Vzorec: O₃Se
MW: 126.96
Bod tání: 117.85…118.85 °C
Hustota: 3.6 g/cm³ (20 °C)
CAS číslo: 13768-86-0
EINECS: 237-385-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR