Tisk…
Ammonium hexafluorotitanate ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
Varování
Synonyms: Ammonium fluotitanate, Ammonium hexafluorotitanate, Ammonium hexafluorotitanate(IV), Ammonium hexafluorotitanate (IV)
43192.22 43192.14 43192.36
43192.22EA 13460 CZK
43192.22 43192.14 43192.36
Ammonium hexafluorotitanate ≥99.99% (metals basis)
Ammonium hexafluorotitanate
Purity: 99.99% CAS No.: 16962-40-6 Molecular Formula: (NH4)2TiF6 Formula Weight: 197.95 Physical Form: Crystalline Appearance: White MDL No.: MFCD00010888
Vzorec: H₈F₆N₂Ti
MW: 197.95
Storage Temperature: Ambient
MDL Number: MFCD00010888
CAS číslo: 16962-40-6
EINECS: 241-036-9
UN: 3288
ADR: 6.1 III