Hydrogensíran amonný, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Ammonium bisulphate
M-Clarity™ quality level = MQ200
455849-100G
455849-25G
SIAL455849-100GEA
6040
CZK
SIAL455849-100G
SIAL455849-25G
Hydrogensíran amonný, Sigma-Aldrich®
Hydrogensíran amonný
Vzorec:
H₅NO₄S Bod varu: 350 °C (1013 hPa) Hustota: 1,787 g/cm³ (20 °C) |
MDL Number:
MFCD00051073 CAS číslo: 7803-63-6 EINECS: 232-265-5 UN: 2506 ADR: 8,II Merck Index: 12,00526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...