Hydrogensíran amonný 99,9 Metals basis
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Ammonium bisulphate
14485.14
14485.22
14485.36
14485.14EA
1570
CZK
14485.14
14485.22
14485.36
Hydrogensíran amonný 99,9 Metals basis
Hydrogensíran amonný
Purity: 99.9%
CAS No.: 7803-63-6
Molecular Formula: NH4HSO4
Formula Weight: 115.10
Physical Form: Lump
Sensitivity: Hygroscopic
UN No.: 2506
MDL No.: MFCD00051073
Vzorec:
H₅NO₄S MW: 115.11 Bod varu: 350 Bod tání: 146.9 °C Hustota: 1.787 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00051073 CAS číslo: 7803-63-6 EINECS: 232-265-5 UN: 2506 ADR: 8 II Merck Index: 12,00526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...