Tisk…

Dimolybdenan amonný (56,5% Mo)

Dodavatel: Thermo Scientific

Synonyms: Ammonium molybdate (di), Diammonium dimolybdate

12215.30 12215.A1 12215.18
12215.30EA 2730 CZK
12215.30 12215.A1 12215.18
Dimolybdenan amonný (56,5% Mo)
Dimolybdenan amonný

CAS No.: 27546-07-2 Molecular Formula: (NH4)2Mo2O7 Formula Weight: 339.95 Physical Form: Powder MDL No.: MFCD00010882

Vzorec: H₈Mo₂N₂O₇
MW: 339,95 g/mol
Hustota: 2,498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010882
CAS číslo: 27546-07-2
EINECS: 248-517-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR