Triphenylphosphine oxide ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12455.14
A12455.22
A12455.36
A12455.14EA
917
CZK
A12455.14
A12455.22
A12455.36
Triphenylphosphine oxide ≥99%
Triphenylphosphine oxide
Purity: 99%
CAS No.: 791-28-6
Molecular Formula: C18H15OP
Formula Weight: 278.29
Flash Point: 180°(356°F)
Melting Point: 153-159°
Boiling Point: 360°
Beilstein Reference: 745854
MDL No.: MFCD00002080
Vzorec:
C₁₈H₁₅OP MW: 278.29 Bod varu: 360 °C Bod tání: 153…159 °C Hustota: 1.200 Bod vzplanutí: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002080 CAS číslo: 791-28-6 EINECS: 212-338-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...