Tisk…

Acetylferrocene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

106860-25G 106860-5G
SIAL106860-25GEA 6640 CZK
SIAL106860-25G SIAL106860-5G
Acetylferrocene, Sigma-Aldrich®
Acetylferrocene
Vzorec: C₁₂H₁₂FeO
Bod tání: 84…85 °C
Hustota: 1 g/cm³ (20 °C)
MDL Number: MFCD00001432
CAS číslo: 1271-55-2
EINECS: 215-043-2
UN: 3467
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR