Tisk…

Tetrabutylammonium hexafluorophosphate, Supelco®

Dodavatel: Merck

Synonyms: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

M-Clarity™ quality level = MQ100

86879-100G 86879-25G
86879-100GEA 20380 CZK
86879-100G 86879-25G
Tetrabutylammonium hexafluorophosphate, Supelco®
Tetrabutylammonium hexafluorophosphate
Vzorec: C₁₆H₃₆F₆NP
MW: 387,43 g/mol
MDL Number: MFCD00011748
CAS číslo: 3109-63-5
EINECS: 221-472-6
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61a6b8d3ef60bb5119538704653475d6ba5ec238&vid=20e9eafeb25c4c542d3bb6bd54a9a073d303481c&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR