Tisk…

Tetrabutylammonium hexafluorophosphate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

M-Clarity™ quality level = MQ200

86874-100G 86874-25G
SIAL86874-100GEA 5060 CZK
SIAL86874-100G SIAL86874-25G
Tetrabutylammonium hexafluorophosphate, Sigma-Aldrich®
Tetrabutylammonium hexafluorophosphate
Vzorec: C₁₆H₃₆F₆NP
MDL Number: MFCD00011748
CAS číslo: 3109-63-5
EINECS: 221-472-6
UN: 3263
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR