Tetrahexylammonium perchlorate
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Tetra-n-hexylammonium perchlorate
30830.06
30830.14
30830.22
30830.06EA
1420
CZK
30830.06
30830.14
30830.22
Tetrahexylammonium perchlorate
Tetrahexylammonium perchlorate
CAS No.: 4656-81-9
Molecular Formula: [CH3(CH2)5]4NClO4
Formula Weight: 454.13
Physical Form: Crystalline
UN No.: 1479
MDL No.: MFCD00043185
Vzorec:
C₂₄H₅₂ClNO₄ MW: 454,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043185 CAS číslo: 4656-81-9 EINECS: 225-095-8 UN: 1479 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...