Tisk…

Tetrapropylammonium chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Tetra-n-propylammonium chloride

M-Clarity™ quality level = MQ200

438243-10G 438243-50G
SIAL438243-10GEA 1350 CZK
SIAL438243-10G SIAL438243-50G
Tetrapropylammonium chloride, Sigma-Aldrich®
Tetrapropylammonium chloride
Vzorec: C₁₂H₂₈ClN
Bod tání: 230 °C
Hustota: 1,0334 g/cm³ (3 °C)
MDL Number: MFCD00038729
CAS číslo: 5810-42-4
EINECS: 227-375-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR