Tisk…

Tetrapropylammonium chloride ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Tetra-n-propylammonium chloride

19379.06 19379.14
19379.06EA 1830 CZK
19379.06 19379.14
Tetrapropylammonium chloride ≥99%
Tetrapropylammonium chloride

Purity: 99+% CAS No.: 5810-42-4 Molecular Formula: C12H28ClN Formula Weight: 221.87 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 240-242° Solubility: Soluble in water, acetone MDL No.: MFCD00038729

Vzorec: C₁₂H₂₈ClN
Bod tání: 240…242 °C
Hustota: 1,0334 g/cm³ (3 °C)
Storage Temperature: Ambient
MDL Number: MFCD00038729
CAS číslo: 5810-42-4
EINECS: 227-375-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR