Tisk…

Cetrimonium chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC

52366-10G 52366-50G
SIAL52366-10GEA 4210 CZK
SIAL52366-10G SIAL52366-50G
Cetrimonium chloride, Sigma-Aldrich®
Cetrimonium chloride
Vzorec: C₁₉H₄₂ClN
Bod tání: 232…233 °C
Hustota: 0,9 g/cm³ (20 °C)
MDL Number: MFCD00011773
CAS číslo: 112-02-7
EINECS: 203-928-6
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR