Tisk…

Cetrimonium chloride ≥96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC

41861.06 41861.14 41861.22 41861.36
41861.06EA 1360 CZK
41861.06 41861.14 41861.22 41861.36
Cetrimonium chloride ≥96%
Cetrimonium chloride

Purity: 96% CAS No.: 112-02-7 Molecular Formula: C19H42ClN Formula Weight: 320.00 Physical Form: Powder Density: 0.968 Appearance: Off-white Odour: Odourless Melting Point: 234° Beilstein Reference: 3657974 UN No.: 1759 MDL No.: MFCD00011773

Vzorec: C₁₉H₄₂ClN
Bod tání: 234 °C
Hustota: 0,968
Storage Temperature: Ambient
MDL Number: MFCD00011773
CAS číslo: 112-02-7
EINECS: 203-928-6
UN: 2923
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR