Tisk…

Dodecyltrimethylammonium chloride, Sigma-Aldrich®

Dodavatel: Merck
Varování

44242-100G 44242-25G
SIAL44242-100GEA 10670 CZK
SIAL44242-100G SIAL44242-25G
Dodecyltrimethylammonium chloride, Sigma-Aldrich®
Dodecyltrimethylammonium chloride
Vzorec: C₁₅H₃₄ClN
MDL Number: MFCD00041974
CAS číslo: 112-00-5
EINECS: 203-927-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR