Tisk…

Dodecyltrimethylammonium bromide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

D5047-100G D5047-25G D5047-5G
SIALD5047-100GEA 63150 CZK
SIALD5047-100G SIALD5047-25G SIALD5047-5G
Dodecyltrimethylammonium bromide, Sigma-Aldrich®
Dodecyltrimethylammonium bromide
Vzorec: C₁₅H₃₄BrN
Bod tání: 264…266 °C
MDL Number: MFCD00011767
CAS číslo: 1119-94-4
EINECS: 214-290-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR