Tisk…

Tetrabutylammonium tetrafluoroborate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A16688.06 A16688.22 A16688.14
A16688.06EA 1350 CZK
A16688.06 A16688.22 A16688.14
Tetrabutylammonium tetrafluoroborate ≥99%
Tetrabutylammonium tetrafluoroborate
Vzorec: C₁₆H₃₆BF₄N
Bod tání: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00011634
CAS číslo: 429-42-5
EINECS: 207-058-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR