Tisk…

Tributylphosphine oxide, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Tri-n-butylphosphineoxide, Oseltamivir impurity H

M-Clarity™ quality level = MQ100

172766-25G 172766-5G
SIAL172766-25GEA 4600 CZK
SIAL172766-25G SIAL172766-5G
Tributylphosphine oxide, Sigma-Aldrich®
Tributylphosphine oxide
Vzorec: C₁₂H₂₇OP
MW: 218,32 g/mol
Bod varu: 348 °C (1013 hPa)
Bod tání: 62…63 °C
MDL Number: MFCD00002082
CAS číslo: 814-29-9
EINECS: 212-394-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR