Bismuth(III) nitrate oxide ≥99.998% (metals basis), Puratronic®
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Bismuth (III) oxide nitrate, (Nitrooxy)oxobismuthine, Bismuth (III) oxynitrate
11122.09
11122.18
11122.09EA
5260
CZK
11122.09
11122.18
Bismuth(III) nitrate oxide ≥99.998% (metals basis), Puratronic®
Bismuth(III) nitrate oxide
Purity: 99.998%
CAS No.: 10361-46-3
Molecular Formula: BiONO3
Formula Weight: 286.99
Physical Form: Powder
UN No.: 1477
MDL No.: MFCD00064844
Vzorec:
BiNO₄ MW: 286,98 g/mol Hustota: 4,93 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064844 CAS číslo: 10361-46-3 EINECS: 233-792-3 UN: 1477 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...