Tisk…

Chlorid-oxid vizmutitý ≥99.95% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: Bismuth oxychloride, Bismuth (III) oxychloride, Bismuth(III) chloride oxide

17131.14 17131.22
17131.14EA 3470 CZK
17131.14 17131.22
Chlorid-oxid vizmutitý ≥99.95% (metals basis)
Chlorid-oxid vizmutitý

Purity: 99.95% CAS No.: 7787-59-9 Molecular Formula: BiClO Formula Weight: 260.43 Physical Form: Powder Density: 7.72 Refractive Index: 2.15 Appearance: Very pale brown Odour: Odourless Melting Point: dec. Merck Reference: 14,1263 MDL No.: MFCD00010895

Vzorec: BiClO
MW: 260,43 g/mol
Bod tání: Dec.
Hustota: 7,72
Storage Temperature: Ambient
MDL Number: MFCD00010895
CAS číslo: 7787-59-9
EINECS: 232-122-7
Merck Index: 14,1263

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR