Tisk…

Diindium trisulphate, anhydrous ≥99.99% (metals basis)

Dodavatel: Thermo Scientific

Synonyms: Diindium tris(sulphate), Indium(III) sulphate, Indium (III) sulphate

40102.14 40102.22
40102.14EA 5360 CZK
40102.14 40102.22
Diindium trisulphate, anhydrous ≥99.99% (metals basis)
Diindium trisulphate

Purity: 99.99% CAS No.: 13464-82-9 Molecular Formula: In2(SO4)3 Formula Weight: 517.83 Physical Form: Solid Density: 3.438 Appearance: Pale grey Odour: Odourless MDL No.: MFCD00011059

Vzorec: O₁₂In₂S₃
MW: 517,83 g/mol
Bod tání: 585 °C
Hustota: 3,438
Storage Temperature: Ambient
MDL Number: MFCD00011059
CAS číslo: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR