Tisk…

Aluminium sulphate hydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Aluminum(III) sulphate xhydrate

M-Clarity™ quality level = MQ200

A0843-250G A0843-500G
SIALA0843-250GEA 8250 CZK
SIALA0843-250G SIALA0843-500G
Aluminium sulphate hydrate, Sigma-Aldrich®
Aluminium sulphate hydrate
Vzorec: Al₂(SO₄)₃·xH₂O
MDL Number: MFCD00149136
CAS číslo: 17927-65-0
EINECS: 233-135-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR