Aluminium sulphate hydrate 98.0-102% ACS
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aluminum(III) sulphate xhydrate
36294.30
36294.A1
36294.30EA
1620
CZK
36294.30
36294.A1
Aluminium sulphate hydrate 98.0-102% ACS
Aluminium sulphate hydrate
Purity: 98.0-102%
CAS No.: 17927-65-0
Molecular Formula: Al2(SO4)3.xH2O (x≈14-18)
Formula Weight: 342.15(anhy)
Physical Form: Crystalline
Density: 1.61
pH: 3.5
Merck Reference: 14,366
MDL No.: MFCD00149136
Vzorec:
Al₂(SO₄)₃·xH₂O MW: 360.17 Hustota: 1.61 Storage Temperature: Ambient |
MDL Number:
MFCD00149136 CAS číslo: 17927-65-0 EINECS: 233-135-0 Merck Index: 14,366 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...