Tisk…

Digallium triselenide ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Gallium (III) selenide

45572.03 45572.06
45572.03EA 2360 CZK
45572.03 45572.06
Digallium triselenide ≥99.99% (metals basis)
Digallium triselenide

Purity: 99.99% CAS No.: 12024-24-7 Molecular Formula: Ga2Se3 Formula Weight: 376.33 Physical Form: 3-12mm Sintered Lump UN No.: 3077 MDL No.: MFCD00016103

Vzorec: Ga₂Se₃
Bod tání: 1010…1030 °C
Hustota: 4,92 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016103
CAS číslo: 12024-24-7
EINECS: 234-693-8
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR