Tisk…

Gallium antimonide ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

12933.03 12933.06
12933.03EA 2510 CZK
12933.03 12933.06
Gallium antimonide ≥99.99% (metals basis)
Gallium antimonide

Purity: 99.99% CAS No.: 12064-03-8 Molecular Formula: GaSb Formula Weight: 191.47 Physical Form: 6mm & down Pieces Odour: Odourless UN No.: 1549 MDL No.: MFCD00016101

Vzorec: GaSb
Bod tání: 710 °C
Hustota: 5,5…5.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016101
CAS číslo: 12064-03-8
EINECS: 235-058-8
UN: 1549
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR