Digallium trisulphide ≥99.999% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Gallium (III) sulphide
41538.03
41538.06
41538.03EA
2970
CZK
41538.03
41538.06
Digallium trisulphide ≥99.999% (metals basis)
Digallium trisulphide
Purity: 99.999%
CAS No.: 12024-22-5
Molecular Formula: Ga2S3
Formula Weight: 235.63
Physical Form: Powder
Density: 3.77
Melting Point: 1255°
UN No.: 3260
MDL No.: MFCD00016106
Vzorec:
Ga₂S₃ MW: 235.64 Bod tání: 1255 °C Hustota: 3.77 Storage Temperature: Ambient |
MDL Number:
MFCD00016106 CAS číslo: 12024-22-5 EINECS: 234-692-2 UN: 3260 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...