Indium phosphide ≥99.999% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Indium(III) phosphide, Indium phosphide
36283.03
36283.06
36283.03EA
6560
CZK
36283.03
36283.06
Indium phosphide ≥99.999% (metals basis)
Indium phosphide
Purity: 99.999%
CAS No.: 22398-80-7
Molecular Formula: InP
Formula Weight: 145.79
Physical Form: 6mm & Down
Density: 4.81
Melting Point: 1070°
Merck Reference: 14,4953
MDL No.: MFCD00016153
Vzorec:
InP Bod tání: 1070 °C Hustota: 4,81 Storage Temperature: Ambient |
MDL Number:
MFCD00016153 CAS číslo: 22398-80-7 EINECS: 244-959-5 Merck Index: 14,4953 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...