Tisk…

Indium phosphide ≥99.9999% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: Indium(III) phosphide, Indium phosphide

13974.06 13974.03
13974.06EA 30200 CZK
13974.06 13974.03
Indium phosphide ≥99.9999% (metals basis)
Indium phosphide

Purity: 99.9999% CAS No.: 22398-80-7 Molecular Formula: InP Formula Weight: 145.79 Physical Form: Pieces Density: 4.81 Melting Point: 1070° Merck Reference: 14,4953 MDL No.: MFCD00016153

Vzorec: InP
MW: 145,79 g/mol
Bod tání: 1070 °C
Hustota: 4,81
Storage Temperature: Ambient
MDL Number: MFCD00016153
CAS číslo: 22398-80-7
EINECS: 244-959-5
Merck Index: 14,4953

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR