Gallium phosphide ≥99.999% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
12266.04
12266.09
12266.18
12266.04EA
5430
CZK
12266.04
12266.09
12266.18
Gallium phosphide ≥99.999% (metals basis)
Gallium phosphide
Purity: 99.999%
CAS No.: 12063-98-8
Molecular Formula: GaP
Formula Weight: 100.69
Physical Form: 6mm & Down Pieces
Density: 4.13
Appearance: Pale orange
Melting Point: 1465°
Merck Reference: 14,4353
MDL No.: MFCD00016109
In semiconducting devices
Vzorec:
GaP Bod tání: 1465 °C Hustota: 4,13 Storage Temperature: Ambient |
MDL Number:
MFCD00016109 CAS číslo: 12063-98-8 EINECS: 235-057-2 Merck Index: 14,4353 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...