Gallium(III) acetylacetonate ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Tris(2,4-pentanedionato-O,O')gallium (III), Gallium (III) 2,4-pentanedionate, Gallium (III) tris(2,4-pentanedionato-O,O'), Gallium(III) Acetylacetonate, Acetylacetone Gallium(III) salt, Tris(2,4-pentanedionato)gallium(III), Ga(acac)3
42086.06
42086.14
42086.06EA
3880
CZK
42086.06
42086.14
Gallium(III) acetylacetonate ≥99.99% (metals basis)
Gallium(III) acetylacetonate
Purity: 99.99%
CAS No.: 14405-43-7
Molecular Formula: C15H21O6Ga
Formula Weight: 367.05
Physical Form: Powder
Density: 1.42
Melting Point: 196-198° dec.
MDL No.: MFCD00013492
Vzorec:
C₁₅H₂₁GaO₆ Hustota: 1,42 Storage Temperature: Ambient |
MDL Number:
MFCD00013492 CAS číslo: 14405-43-7 EINECS: 238-377-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...