Tisk…

Digallium trioxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

215066-10G 215066-50G
SIAL215066-10GEA 6940 CZK
SIAL215066-10G SIAL215066-50G
Digallium trioxide, Sigma-Aldrich®
Digallium trioxide
Vzorec: Ga₂O₃
Hustota: 5,95 g/cm³ (27 °C)
MDL Number: MFCD00011020
CAS číslo: 12024-21-4
EINECS: 234-691-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR