Indium(I) iodide, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
413658-1G
413658-5G
SIAL413658-1GEA
3010
CZK
SIAL413658-1G
SIAL413658-5G
Indium(I) iodide, Sigma-Aldrich®
Indium(I) iodide
Vzorec:
InI MW: 241,72 g/mol Bod varu: 711 °C (1013 hPa) Bod tání: 351 °C Hustota: 5,32 g/cm³ (20 °C) |
MDL Number:
MFCD00135543 CAS číslo: 13966-94-4 EINECS: 237-746-3 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...