Tisk…

Indium(I) iodide, ultra dry ≥99.998% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Ampuled under argon

35700.06 35700.03
35700.06EA 14590 CZK
35700.06 35700.03
Indium(I) iodide, ultra dry ≥99.998% (metals basis)
Indium(I) iodide

Purity: 99.998% CAS No.: 13966-94-4 Molecular Formula: InI Formula Weight: 241.72 Physical Form: -10 Mesh Beads, Ampouled under argon Sensitivity: Moisture and Light Sensitive MDL No.: MFCD00135543

Vzorec: InI
Bod varu: 711 °C (1013 hPa)
Bod tání: 351 °C
Hustota: 5,32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135543
CAS číslo: 13966-94-4
EINECS: 237-746-3
UN: 3260
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR