Strontium titanate ≥99% (metals basis)
Dodavatel: Thermo Fisher Scientific
11399.30
11399.A1
11399.30EA
2030
CZK
11399.30
11399.A1
Strontium titanate ≥99% (metals basis)
Strontium titanate
Purity: 99+%
CAS No.: 12060-59-2
Molecular Formula: SrTiO3
Formula Weight: 183.51
Physical Form: Powder
Density: 4.810
Melting Point: 2060°
Solubility: Insoluble in water and most solvents
MDL No.: MFCD00049554
Used in electronics and in electrical insulation
Vzorec:
O₃SrTi MW: 183.5 Bod tání: 2060 °C Hustota: 4.810 Storage Temperature: Ambient |
MDL Number:
MFCD00049554 CAS číslo: 12060-59-2 EINECS: 235-044-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...