Barium titanate ≥99.7% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide
88267.22
88267.36
88267.22EA
3160
CZK
88267.22
88267.36
Barium titanate ≥99.7% (metals basis)
Barium titanate
Purity: 99.7%
CAS No.: 12047-27-7
Molecular Formula: BaTiO3
Formula Weight: 233.19
Physical Form:
Vzorec:
BaO₃Ti MW: 233,21 g/mol Bod tání: 1620 °C Hustota: 5,85 Storage Temperature: Ambient |
MDL Number:
MFCD00003447 CAS číslo: 12047-27-7 EINECS: 234-975-0 UN: 1564 ADR: 6.1,III Merck Index: 14,1000 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...