Tisk…

Barium titanate ≥99.95% (metals basis)

Dodavatel: Thermo Scientific
Varování

Synonyms: Barium titanate, Barium tantalum oxide, Barium titanate (IV), Barium titanium oxide

45083.14 45083.06
45083.14EA 13760 CZK
45083.14 45083.06
Barium titanate ≥99.95% (metals basis)
Barium titanate

Purity: 99.95% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: 3-6mm Pieces Odour: Odourless Merck Reference: 14,1000 MDL No.: MFCD00003447

Vzorec: BaO₃Ti
MW: 233,21 g/mol
Bod tání: 1620 °C
Hustota: 5,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS číslo: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR