Tisk…

Barium titanate ≥99.99% (metals basis), Puratronic®

Dodavatel: Thermo Fisher Scientific
Varování

10652.14 10652.22
10652.14EA 4260 CZK
10652.14 10652.22
Barium titanate ≥99.99% (metals basis), Puratronic®
Barium titanate

Purity: 99.99% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: Powder Density: 5.85 Appearance: White Odour: Odourless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447

Vzorec: BaO₃Ti
Bod tání: 1620 °C
Hustota: 5,85
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS číslo: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR