Tisk…

Calcium iodide, ultra dry ≥99.999% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování


Ampuled under argon

44254.14 44254.22
44254.14EA 7180 CZK
44254.14 44254.22
Calcium iodide, ultra dry ≥99.999% (metals basis)
Calcium iodide

Purity: 99.999% CAS No.: 10102-68-8 Molecular Formula: CaI2 Formula Weight: 293.89 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.956 Appearance: White to pale yellow Odour: Odourless Melting Point: 784° Boiling Point: 1100° Merck Reference: 14,1677 MDL No.: MFCD00010910

Vzorec: CaI₂
Bod varu: 1100 °C
Bod tání: 784 °C
Hustota: 3,956
Storage Temperature: Ambient
MDL Number: MFCD00010910
CAS číslo: 10102-68-8
EINECS: 233-276-8
Merck Index: 14,1677

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR