Calcium iodide, ultra dry ≥99.99% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
Ampuled under argon
43674.22
43674.14
43674.36
43674.22EA
19630
CZK
43674.22
43674.14
43674.36
Calcium iodide, ultra dry ≥99.99% (metals basis)
Calcium iodide
Purity: 99.99%
CAS No.: 10102-68-8
Molecular Formula: CaI2
Formula Weight: 293.89
Physical Form: Powder, Ampouled under argon
Density: 3.956
Appearance: White to pale yellow
Odour: Odourless
Melting Point: 784°
Boiling Point: 1100°
Merck Reference: 14,1677
MDL No.: MFCD00010910
Vzorec:
CaI₂ Bod varu: 1100 °C Bod tání: 784 °C Hustota: 3,956 Storage Temperature: Ambient |
MDL Number:
MFCD00010910 CAS číslo: 10102-68-8 EINECS: 233-276-8 Merck Index: 14,1677 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...