Tisk…

Karbid vápenatý

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Calcium acetylide

14308.A1 14308.D9
14308.A1EA 2700 CZK
14308.A1 14308.D9
Karbid vápenatý
Karbid vápenatý

CAS No.: 75-20-7 Molecular Formula: C2Ca Formula Weight: 64.09 Physical Form: 9-40mm Lump Density: 2.22 Sensitivity: Moisture Sensitive Appearance: Greyish black Melting Point: 2300° Beilstein Reference: 3909011 Merck Reference: 14,1656 UN No.: 1402 MDL No.: MFCD00010905

Vzorec: CaC₂
MW: 64.1
Bod tání: 2300
Hustota: 2.22
Storage Temperature: Ambient
MDL Number: MFCD00010905
CAS číslo: 75-20-7
EINECS: 200-848-3
UN: 1402
ADR: 4.3 II
Merck Index: 12,01696

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR